Learn More
Polysciences, Inc. Polybead Carboxylate Microspheres 3.00 um, 5 mL
SDP
Catalog No. NC1172814
Encompass
Polybead Carboxylate Microspheres are monodisperse polystyrene microspheres that contain surface carboxyl groups. This allows for the covalent binding of proteins to the bead surface. These microspheres are packaged as 2.5% solids (w/v) aqueous suspensions.
Catalog No. NC1172814 Supplier Polysciences, Inc. Supplier No. 098505
Polybead Carboxylate Microspheres are monodisperse polystyrene microspheres that contain surface carboxyl groups. This allows for the covalent binding of proteins to the bead surface. These microspheres are packaged as 2.5% solids (w/v) aqueous suspensions.