Learn More
Polysciences, Inc. Polybead® Carboxylate Sampler Kit I
SDP
Catalog No. NC3876577
Encompass
Kit contains 5ml of carboxylate polystyrene monodisperse microspheres (2.5% solids in water) for each of these nominal sizes: 0.5µm, 0.75µm, 1.0µm, 2.0µm and 3.0µm.
Catalog No. NC3876577 Supplier Polysciences, Inc. Supplier No. 198191
Kit contains 5ml of carboxylate polystyrene monodisperse microspheres (2.5% solids in water) for each of these nominal sizes: 0.5µm, 0.75µm, 1.0µm, 2.0µm and 3.0µm.