Learn More
Polysciences, Inc. Polybead Microspheres 90.00um, 5ml
SDP
Catalog No. NC1458110
Encompass
Polybead Microspheres 90.00m, 5mlPolybead Microspheres are monodisperse polystyrene microspheres available in sizes from 50nm to 90m. Diameters 4.5m and larger are crosslinked with divinylbenzene (DVB). Our experienced chemists control the synthesis to provide precise monodisperse particle size distributions. These particles contain a slight anionic charge from sulfate ester.Most Polybead Microspheres are packaged in an economical 2.5% solids (w/v) aqueous suspension with minimal surfactant in the final preparation. Polybead Microspheres are ideally suited for protein binding using passive adsorption techniques.
Catalog No. NC1458110 Supplier Polysciences, Inc. Supplier No. 073155
Polybead Microspheres 90.00m, 5mlPolybead Microspheres are monodisperse polystyrene microspheres available in sizes from 50nm to 90m. Diameters 4.5m and larger are crosslinked with divinylbenzene (DVB). Our experienced chemists control the synthesis to provide precise monodisperse particle size distributions. These particles contain a slight anionic charge from sulfate ester.Most Polybead Microspheres are packaged in an economical 2.5% solids (w/v) aqueous suspension with minimal surfactant in the final preparation. Polybead Microspheres are ideally suited for protein binding using passive adsorption techniques.