Learn More
Santa Cruz Biotechnology POLYBRENE(10MG/ML)1ML
Catalog No. NC9840454
Encompass
1 ml; 10mg/ml in sterile water -highly efficient infection reagent used to introduce retroviral vectors into mammalian cells -also known as hexamethrine bromide, 1,5-Dimethyl-1,5-diazaundecamethylene polymethobromide-Polybrene is a registered trademark of Abbott Laboratories Corp.-Store at -20 deg. C. 10 mg/ml -Molecular Formula: C13H30Br2N2-Molecular Weight: 374.1987 [g/mol]
Catalog No. NC9840454 Supplier Santa Cruz Biotechnology Supplier No. SC134220
Add to Cart
Add to Cart
1 ml; 10mg/ml in sterile water -highly efficient infection reagent used to introduce retroviral vectors into mammalian cells -also known as hexamethrine bromide, 1,5-Dimethyl-1,5-diazaundecamethylene polymethobromide-Polybrene is a registered trademark of Abbott Laboratories Corp.-Store at -20 deg. C. 10 mg/ml -Molecular Formula: C13H30Br2N2-Molecular Weight: 374.1987 [g/mol]