Learn More
Sigma Aldrich Fine Chemicals Biosciences Hexadimethrine bromide, 28728-55-4, MFCD00133397, 10 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 10768910G
Encompass
Antiheparin agent. A 3mg quantity has been found to enhance peptide degradation in automated sequential analysis. Store: 2-8°C, 95%, Synonym: 1,5-Dimethyl-1,5-diazaundecamethylene polymethobromide, Polybrene.
Catalog No. NC0663391