Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Polybrene Infection/Transfection Reagent

Catalog No. TR1003G Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TR1003G 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TR1003G Supplier MilliporeSigma™ Supplier No. TR1003G
Only null left
Add to Cart
Add to Cart

A highly efficient method of gene transfer into mammalian cells leveraging infection with retroviral vectors.

The efficiency of retroviral infection is enhanced significantly, 100 to 1000 fold in some cells, by including polybrene during the infection. Polybrene with DMSO shock is also used to mediate DNA transfer into a variety of cell types, such as: CHO, chicken embryo fibroblasts, NIH3T3 cells, and myeloid cells.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C up to 2 years
Description Presentation: 10mg polybrene per mL sterile Ultra Pure water.
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.