Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polybutenes

Catalog No. 3887261L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-872-61L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-872-61L Supplier Sigma Aldrich Supplier No. 3887261L
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hydrophobic material is derivatized to make detergents and anti-rust additives. Adds tack, flexibility and water repellency to adhesives and coatings. Contributes 'cling' to LLDPE films. Plasticizer for polymers, lubricants and metal-working fluids.remainder 1- and 2-butenes

Specifications

CAS 9003-29-6
Density 0.908 g/mL (at 25°C)
Linear Formula [-CH2C(CH3)2-]x[-CH2CH(C2H5)-]y
Refractive Index n20/D 1.504
MDL Number MFCD00197897
Quantity 1 L
Molecular Weight (g/mol) 2300
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.