Learn More
Scientific Polymer Polycaprolactone, 24980-41-4, 250 G
Catalog No. NC9992081
Encompass
Linear Formula: (C6H10O2)n, Synonym: 2-Oxepanone homopolymer, 6-Caprolactone polymer, MP: 60 °C (lit.). Polycaprolactone may find use as an extrusion aid, die lubricant, mold release, pigment and filler dispersion aid and polyester segments in urethanes and block polyesters.
Catalog No. NC9992081 Supplier Scientific Polymer Supplier No. 1030
Linear Formula: (C6H10O2)n, Synonym: 2-Oxepanone homopolymer, 6-Caprolactone polymer, MP: 60 °C (lit.). Polycaprolactone may find use as an extrusion aid, die lubricant, mold release, pigment and filler dispersion aid and polyester segments in urethanes and block polyesters.