Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polycaprolactone diol

Catalog No. 189405250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-940-5250G 250 g
18-940-5500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-940-5250G Supplier Sigma Aldrich Supplier No. 189405250G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Polycaprolactone diol(PCL-diol) may be used as a precursor to biodegradable and bioresorbable polycaprolactone. PCL-diol in combination with polytetrahydrofuran diol (PTHF-diol) and ceric ammonium nitrate may be used to initiate polymerization of methyl methacrylate (MMA).

Specifications

CAS 36890-68-3
Density 1.073 g/mL (at 25°C)
Linear Formula C4H8O3(C6H10O2)n
Quantity 250 g
MDL Number MFCD00084404
Synonym Diethylene glycol-initiated polycaprolactone; Polycaprolactone oxydiethylene ester
Molecular Weight (g/mol) 530
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.