Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polycaprolactone diol

Catalog No. 189421500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-942-1500G 500 g
18-942-1250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-942-1500G Supplier Sigma Aldrich Supplier No. 189421500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Polycaprolactone diol (PCL-diol), along with polytetrahydrofuran diol (PTHF-diol) and ceric ammonium nitrate may be used to polymerize methyl methacrylate (MMA). The product may be used to prepare biocompatible poly(ester urethane)urea (PEUU) and as a precursor for polyurethane (PU) thermoplastic polymer.

Specifications

CAS 36890-68-3
Density 1.071 g/mL (at 25°C)
Linear Formula C4H8O3(C6H10O2)n
Quantity 500 g
MDL Number MFCD00084404
Synonym Diethylene glycol-initiated polycaprolactone; Polycaprolactone oxydiethylene ester
Molecular Weight (g/mol) 2000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.