Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polycaprolactone triol

Catalog No. 200387500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-038-7500G 500 g
20-038-7250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-038-7500G Supplier Sigma Aldrich Supplier No. 200387500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Polycaprolactone triol (PCL-triol) may be used to prepare biodegradable elastomer with citric acid for soft tissue engineering applications. A number of studies report the use of PCL-triol in the preparation of biodegradable polyurethane, which finds a number of uses as biomedical implants.

Specifications

CAS 37625-56-2
Density 1.07 g/mL (at 25°C)
Linear Formula C2H5C[CH2O[CO(CH2)5 O]nH]3
MDL Number MFCD00084405
Quantity 500 g
Molecular Weight (g/mol) 300
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.