Learn More
Beckman Coulter POLYCARB BTTL ASSBLY 6/PK
Catalog No. NC9691797
Encompass_Preferred
Polycarbonate Bottle/Assembly,Quantity:6,Size(mm):38 x 102, Vol.(mL): 70.0
Catalog No. NC9691797 Supplier Beckman Coulter Supplier No. 355622
Polycarbonate Bottle/Assembly,Quantity:6,Size(mm):38 x 102, Vol.(mL): 70.0