Learn More
Mitegen Polycarbonate 100-Cell Storage Box, For Various IC Biomedical Products, Holds Non-Threaded or Internally Threaded Vials, For Cryogenic Storage, Case of 24
SDP
Catalog No. NC3372997
Encompass
For use with the IC Biomedical CXR500, LS3000, LS4800, LS6000, 10K, 24K, 38K, LABS 20K, LABS 38K, LABS 40K, LABS 80K, and LABS 94K products. This Polycarbonate 100-Cell Storage Box is an ideal solution for organized and secure cryogenic sample management in laboratories. Each durable box is precisely designed to hold up to 100 vials, accommodating both non-threaded and internally threaded types, offering versatile storage capabilities. Constructed from robust polycarbonate, these boxes provide excellent thermal stability and resistance to extreme cryogenic temperatures, safeguarding valuable biological materials during long-term storage and transport. The clear material allows for easy visual inventory, streamlining sample retrieval and reducing handling time. This is a case of 24.
Catalog No. NC3372997 Supplier Mitegen Supplier No. TWR24K9C44
For use with the IC Biomedical CXR500, LS3000, LS4800, LS6000, 10K, 24K, 38K, LABS 20K, LABS 38K, LABS 40K, LABS 80K, and LABS 94K products. This Polycarbonate 100-Cell Storage Box is an ideal solution for organized and secure cryogenic sample management in laboratories. Each durable box is precisely designed to hold up to 100 vials, accommodating both non-threaded and internally threaded types, offering versatile storage capabilities. Constructed from robust polycarbonate, these boxes provide excellent thermal stability and resistance to extreme cryogenic temperatures, safeguarding valuable biological materials during long-term storage and transport. The clear material allows for easy visual inventory, streamlining sample retrieval and reducing handling time. This is a case of 24.