Learn More
Beckman Coulter Open-Top Thickwall Polycarbonate Tube, 8 x 51mm, 1 mL, 25/pk
Catalog No. NC9437570
Encompass_Preferred
Thickwall Polycarbonate Tubes for use in Ultracentrifuge Rotors: Type 50.4 Ti (Part Number 347299); Required Tube Adapter.
Catalog No. NC9437570 Supplier Beckman Coulter Supplier No. 355657
Thickwall Polycarbonate Tubes for use in Ultracentrifuge Rotors: Type 50.4 Ti (Part Number 347299); Required Tube Adapter.