Learn More
Beckman Coulter POLYCARB CENTRI TB 10ML25/PK
Catalog No. NC9529245
Encompass_Preferred
Thickwall Polycarbonate Tubes; ,Size(mm):16 x 64, Vol.(mL): 10.0
Catalog No. NC9529245 Supplier Beckman Coulter Supplier No. 355647
Thickwall Polycarbonate Tubes; ,Size(mm):16 x 64, Vol.(mL): 10.0