Learn More
Thomas Scientific POLYCARB CRYO RACK 50 VIALS
SDP
Catalog No. NC2170065
Encompass
This durable and autoclavable Cryo rack can hold 50 vials. Unique design at the bottom of each well lets vials be unscrewed without removing them from the rack. The rack features an alpha numeric index, and anti-skid rubber feet.
This item is not returnable. View return policy
Catalog No. NC2170065 Supplier Thomas Scientific Supplier No. 1160X38
This durable and autoclavable Cryo rack can hold 50 vials. Unique design at the bottom of each well lets vials be unscrewed without removing them from the rack. The rack features an alpha numeric index, and anti-skid rubber feet.