Learn More
Polyscience 28L Polycarbonate Open Tank
SDP
Catalog No. NC1632071
Encompass_Preferred
PolyScience™ 28 Liter Polycarbonate Open Tank85°C Maximum TemperatureWorking Access (LxWxD):20.6 x 12 x 8 in52.3 x 30.5 x 20.3 cmOverall Dimensions (LxWxD):21.1 x 12.75 x 9.13 in56.1 x 32.4 x 23.2 cm
Catalog No. NC1632071 Supplier Polyscience Supplier No. T28PA1
PolyScience™ 28 Liter Polycarbonate Open Tank85°C Maximum TemperatureWorking Access (LxWxD):20.6 x 12 x 8 in52.3 x 30.5 x 20.3 cmOverall Dimensions (LxWxD):21.1 x 12.75 x 9.13 in56.1 x 32.4 x 23.2 cm