Learn More
Alkali Scientific 2" Polycarbonate 25-Cell Freezer box
SDP
Catalog No. 502389353
Encompass_Preferred
Polycarbonate 2" boxes hold most popular cryo and cold storage vials and fit into standard freezer racks. Boxes have clear lids with locator grid to facilitate sample identification. Polycarbonate boxes are designed to withstand temperature extremes of -196 to 121. Boxes have drain holes for venting and safety. Keyed lid prevents misalignment of grid. Dimensions (L x W x H): 3 x 3 x 2 1/16″ (76 x 76 x 53mm), cell opening 0.96 inch.
Catalog No. 50-238-9353 Supplier Alkali Scientific Supplier No. MPB225
Polycarbonate 2" boxes hold most popular cryo and cold storage vials and fit into standard freezer racks. Boxes have clear lids with locator grid to facilitate sample identification. Polycarbonate boxes are designed to withstand temperature extremes of -196 to 121. Boxes have drain holes for venting and safety. Keyed lid prevents misalignment of grid. Dimensions (L x W x H): 3 x 3 x 2 1/16″ (76 x 76 x 53mm), cell opening 0.96 inch.