Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Polycarbonate Face Shield
GREENER_CHOICE

Catalog No. 114095 Shop All Thermo Scientific Products
Change view
Click to view available options
Lens Tint:
Clear
Length (English):
11.6 in.
Width (English):
7.8 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Length (English) Width (English)
11-409-5 Clear 11.6 in. 7.8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-409-5 Supplier Thermo Scientific™ Supplier No. 63550001
  • $447.50
    $308.65 / Each

    Save $138.85

    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,555.00
    $1,073.65 / Case of 4 EA

    Save $481.35

    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Wear this adjustable polycarbonate face shield over safety glasses. It provides impact protection and features an optically clear 1/16 in. thick polycarbonate window.

Wear the adjustable Thermo Scientific™ Nalgene™ Polycarbonate Faceshield over safety glasses. With an optically clear window made of 1/16 in. (1.5mm) thick polycarbonate, this faceshield provides impact protection as well as protection from UV light in the 200-360nm range. It is useful in germicidal procedures as well as reading of chromatographic/electrophoretic strips and gels. Complies with OSHA 29CFR1910.1030 and ANSI Z87.1-1989 standards for use with bloodborne pathogens.

Features:
  • Wraparound design offers both front and side protection while height provides coverage from the bottom of the neck to the top of the head
  • Features a comfortable and adjustable polyethylene headdress
  • Biohazard/Transparent
Note:UV protection from 200 to 360nm (UV radiation may cause the polycarbonate to develop a slight yellow/brown tint.)
Note: Do not use with radioisotopes. Use Nalgene beta-radiation shields when working with beta-radioisotopes. Do not autoclave.

Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in. 

 

 

TRUSTED_SUSTAINABILITY

Specifications

Thickness (Metric) 1.5 mm
Thickness (English) 0.060 in.
Width (English) 7.8 in.
Description Adjustable Face Shield
Lens Tint Clear
Product Type Shield Face PC/PE
Length (English) 11.6 in.
No. per Case 1/Cs.
Material Polycarbonate

Note: Do not use with radioisotopes. Use Nalgene beta-radiation shields when working with beta-radioisotopes. Do not autoclave.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.