Learn More
MSC Polycarbonate Faceshield Window, 3M, Shade 5.0, 0.08 in Thick, 10/Case
Catalog No. 5000782048
Encompass
This polycarbonate face shield window provides protection against impact, splash hazards, and flying debris in industrial environments. Featuring a green tint with Shade 5, it meets ANSI Z87.1 and CSA Z94.3 standards, and blocks up to 97.5% of IR radiation.
  • Made from durable polycarbonate material
  • Green tint provides Shade 5 protection
  • Features 0.08 in. (0.2 cm) thickness for robust protection
  • Molded design for optimal fit
  • Meets ANSI Z87.1 and CSA Z94.3 safety standards
  • Suitable for applications including welding, grinding, and metal working
Catalog No. 50-007-82048 Supplier MSC Supplier No. 00922500
This polycarbonate face shield window provides protection against impact, splash hazards, and flying debris in industrial environments. Featuring a green tint with Shade 5, it meets ANSI Z87.1 and CSA Z94.3 standards, and blocks up to 97.5% of IR radiation.
  • Made from durable polycarbonate material
  • Green tint provides Shade 5 protection
  • Features 0.08 in. (0.2 cm) thickness for robust protection
  • Molded design for optimal fit
  • Meets ANSI Z87.1 and CSA Z94.3 safety standards
  • Suitable for applications including welding, grinding, and metal working