Learn More
MSC POLYCARBONATE FACESHIELD
Catalog No. 5000795212
Encompass
The 3M WP96 faceshield is a robust 1.8 mm thick molded polycarbonate visor designed to offer comprehensive face protection in industrial environments. It safeguards against high-speed particles, molten metal splashes, liquid splashes, and UV radiation when used with compatible headgear.
  • Molded polycarbonate construction for durability
  • Provides high impact and heat protection
  • Meets ANSI Z87.1 and EN 166 standards
  • Features UV protection
  • Designed for easy snap-in replacement
  • Offers optical clarity class 1
  • Protects against liquid splash and molten metal
Catalog No. 50-007-95212 Supplier MSC Supplier No. 92017359
The 3M WP96 faceshield is a robust 1.8 mm thick molded polycarbonate visor designed to offer comprehensive face protection in industrial environments. It safeguards against high-speed particles, molten metal splashes, liquid splashes, and UV radiation when used with compatible headgear.
  • Molded polycarbonate construction for durability
  • Provides high impact and heat protection
  • Meets ANSI Z87.1 and EN 166 standards
  • Features UV protection
  • Designed for easy snap-in replacement
  • Offers optical clarity class 1
  • Protects against liquid splash and molten metal