Learn More
Sterlitech Corporation Polycarbonate (PCTG) Gold-Coated Membrane Filters, 0.8 Micron, 40/20nm Coating, 47mm, 10/Pk
SDP
Catalog No. NC1706421
Encompass_Preferred
POLYCARBONATE (PCTG) GOLD-COATED MEMBRANE FILTERS, 0.8 MICRON, 40/20NM COATING, 47MM, 10/PK
Catalog No. NC1706421 Supplier Sterlitech Corporation Supplier No. 1270008
POLYCARBONATE (PCTG) GOLD-COATED MEMBRANE FILTERS, 0.8 MICRON, 40/20NM COATING, 47MM, 10/PK