Learn More
Braintree Scientific Rodent Irradiator Disk
SDP
Catalog No. 501954973
Encompass
Using multiple Rad Disk, it is possible to conduct sequential irradiations on groups of up to 20 mice (without dividers) without the need of autoclaving cages between each use, because the interior of the Rad Disk maintains a filter protected sterile environment. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4973 Supplier Braintree Scientific Supplier No. IRDPM
Using multiple Rad Disk, it is possible to conduct sequential irradiations on groups of up to 20 mice (without dividers) without the need of autoclaving cages between each use, because the interior of the Rad Disk maintains a filter protected sterile environment. **Sold for research purposes only and not for human use**