Learn More
MSC POLYCARBONATE SHORT VISOR
Catalog No. 5000782709
Encompass
This short visor is molded from polycarbonate, providing impact protection for various applications. It features anti-fog and anti-scratch coatings, designed to fit X5000 Series Safety Helmets.
  • Molded from polycarbonate for impact protection
  • Features an anti-fog coating
  • Features an anti-scratch coating
  • Designed for X5000 Series safety helmets
  • Slide in/out attachment type
  • Clear color
Catalog No. 50-007-82709 Supplier MSC Supplier No. 90259573
This short visor is molded from polycarbonate, providing impact protection for various applications. It features anti-fog and anti-scratch coatings, designed to fit X5000 Series Safety Helmets.
  • Molded from polycarbonate for impact protection
  • Features an anti-fog coating
  • Features an anti-scratch coating
  • Designed for X5000 Series safety helmets
  • Slide in/out attachment type
  • Clear color