Learn More
MSC POLYCARBONATE SHORT VISOR
Catalog No. 5000782741
Encompass
This short visor for the X5000 Safety Helmet is molded from polycarbonate, providing impact protection for various applications. It features anti-fog and anti-scratch coatings for enhanced visibility and durability.
  • Molded from polycarbonate for impact protection
  • Features an anti-fog coating
  • Features a scratch-resistant coating
  • Compatible with X5000 series safety helmets
  • Gray color
  • Slide in/out attachment
Catalog No. 50-007-82741 Supplier MSC Supplier No. 90259581
This short visor for the X5000 Safety Helmet is molded from polycarbonate, providing impact protection for various applications. It features anti-fog and anti-scratch coatings for enhanced visibility and durability.
  • Molded from polycarbonate for impact protection
  • Features an anti-fog coating
  • Features a scratch-resistant coating
  • Compatible with X5000 series safety helmets
  • Gray color
  • Slide in/out attachment