Learn More
MSC POLYCARBONATE SHORT VISOR
Catalog No. 5000765617
Encompass
The 3M™ short visor is molded from polycarbonate for impact protection in various applications. It features an anti-fog and anti-scratch coating and is available in clear, grey, and amber.
  • Molded from polycarbonate for impact protection.
  • Features an anti-fog and anti-scratch coating.
  • Fits 3M™ SecureFit™ X5000 Series Safety Helmets.
Catalog No. 50-007-65617 Supplier MSC Supplier No. 90259565
The 3M™ short visor is molded from polycarbonate for impact protection in various applications. It features an anti-fog and anti-scratch coating and is available in clear, grey, and amber.
  • Molded from polycarbonate for impact protection.
  • Features an anti-fog and anti-scratch coating.
  • Fits 3M™ SecureFit™ X5000 Series Safety Helmets.