Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 500 mL Square Bottle, Polycarbonate, 12/Tray, 24/Case

Catalog No. 01213109 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
01-213-109 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-213-109 Supplier Corning™ Supplier No. 431495
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Square bottle for pharmaceutical quality control and life sciences applications.

  • For pharmaceutical quality control and life sciences applications
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Color Clear
Material Polycarbonate
No. per Case 24
No. per Pack 12
Packaging Shrink Wrapped Tray
Sterility Non-sterile
Closure Included Yes
Graduated Yes
Mouth Wide
Autoclavable Not Autoclavable
Shape Square
Disposable Reusable
Product Type Media Storage Bottle
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.