Learn More
Medchemexpress LLC Polycarbophil | 9003-97-8 | 500 MG
SDP
Catalog No. 5000376737
Encompass_Preferred
Polycarbophil is an orally active hydrophilic polymer that achieves bioadhesion through hydrogen bonding between carboxyl groups and mucosal surfaces. It prolongs compound retention time and regulates compound release through swelling properties, making it effective in relieving constipation and diarrhea.
  • Orally active hydrophilic polymer
  • Achieves bioadhesion through hydrogen bonding
  • Prolongs compound retention time
  • Regulates compound release through swelling properties
  • Relieves constipation
  • Relieves diarrhea
Catalog No. 50-003-76737 Supplier Medchemexpress LLC Supplier No. HY154659500MG
Polycarbophil is an orally active hydrophilic polymer that achieves bioadhesion through hydrogen bonding between carboxyl groups and mucosal surfaces. It prolongs compound retention time and regulates compound release through swelling properties, making it effective in relieving constipation and diarrhea.
  • Orally active hydrophilic polymer
  • Achieves bioadhesion through hydrogen bonding
  • Prolongs compound retention time
  • Regulates compound release through swelling properties
  • Relieves constipation
  • Relieves diarrhea