Learn More
Polylc Inc PolyLC PolyCAT A Column, Weak Cation-Exchange, High Resolution, 1.0 mm x 150 mm
SDP
Catalog No. NC2097702
Encompass
The PolyLC PolyCAT A Column offers high-performance weak cation-exchange chromatography for biomolecule analysis, including proteins and antibodies. Engineered with a poly(aspartic acid) stationary phase, it ensures sharp resolution and high recovery, making it ideal for diverse research applications. Its superior selectivity for proteins with a pI above 6.0 sets it apart in biopharmaceutical and proteomics research.
This item is not returnable. View return policy
Catalog No. NC2097702 Supplier Polylc Inc Supplier No. 151CT0510
The PolyLC PolyCAT A Column offers high-performance weak cation-exchange chromatography for biomolecule analysis, including proteins and antibodies. Engineered with a poly(aspartic acid) stationary phase, it ensures sharp resolution and high recovery, making it ideal for diverse research applications. Its superior selectivity for proteins with a pI above 6.0 sets it apart in biopharmaceutical and proteomics research.