Learn More
Crescent Chemical Co Inc POLYCLAR AT PRACT.
SDP
Supplier:  Crescent Chemical Co Inc SE33162.02
Encompass_Preferred
Polyclar AT pract. Water-insoluble polyvinylpyrrolidone for binding phenols. Size - 500G Storage Conditions - +15 C TO +30 C Catalog Number - 33162.02 CAS 9003-39-8
Catalog No. 50-248-041