Learn More
Polyscience polyclear MIX 30 PLUS (Case)
SDP
Catalog No. NC1653078
Encompass_Preferred
PolyScience™ polyclear MIX 30 PLUS (case)15 to 90° Temperature Rangecase = 5x1/2 gallon (1.9 L)General purpose fluid for routine applications above 15°C, prevents algae growth and premature rust formation.
Catalog No. NC1653078 Supplier Polyscience Supplier No. 004300063
PolyScience™ polyclear MIX 30 PLUS (case)15 to 90° Temperature Rangecase = 5x1/2 gallon (1.9 L)General purpose fluid for routine applications above 15°C, prevents algae growth and premature rust formation.