Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Presenilin-2 Polyclonal Antibody, Rabbit Host, Unconjugated, 100 μL
SDP

Catalog No. 50194927
Encompass

Presenilin 2 Antibody is a rabbit polyclonal antibody designed to detect endogenous levels of the carboxy-terminal fragment (23 kDa) and the full-length (54 kDa) human presenilin 2 protein. This antibody does not cross-react with endogenous presenilin 1 and is validated for western blotting and immunoprecipitation applications.

  • Host species: rabbit
  • Species reactivity: human, mouse, rat, monkey
  • Purified by protein A and peptide affinity chromatography
  • Immunized with a synthetic peptide corresponding to residues surrounding amino acid 330 of human presenilin 2
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA and 50% glycerol

Catalog No. 50-194-927 Supplier Cell Signaling Technology Supplier No. 2192S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Presenilin 2 Antibody is a rabbit polyclonal antibody designed to detect endogenous levels of the carboxy-terminal fragment (23 kDa) and the full-length (54 kDa) human presenilin 2 protein. This antibody does not cross-react with endogenous presenilin 1 and is validated for western blotting and immunoprecipitation applications.

  • Host species: rabbit
  • Species reactivity: human, mouse, rat, monkey
  • Purified by protein A and peptide affinity chromatography
  • Immunized with a synthetic peptide corresponding to residues surrounding amino acid 330 of human presenilin 2
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA and 50% glycerol

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.