Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology STAG2 Polyclonal Antibody, Rabbit Host, 100 μL
SDP

Catalog No. 50195274
Encompass

This polyclonal antibody detects endogenous levels of total STAG2 protein. The product is produced by immunizing rabbits with a synthetic peptide corresponding to residues surrounding Glu31 of human STAG2. The antibody is purified via protein A and peptide affinity chromatography and is supplied in a formulation containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, and 50% glycerol.

  • Validated for Western blotting and immunoprecipitation applications.
  • Species reactivity includes human, mouse, rat, and monkey.
  • Target specificity ensures no cross-reactivity with other STAG proteins such as STAG1 or STAG3.
  • Supplied at a concentration of 49.3 μg/mL.
  • Recommended dilution for Western blotting is 1:1,000 and 1:100 for immunoprecipitation.
  • Storage at −20°C is required; do not aliquot.

This item is not returnable. View return policy
Catalog No. 50-195-274 Supplier Cell Signaling Technology Supplier No. 4239S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This polyclonal antibody detects endogenous levels of total STAG2 protein. The product is produced by immunizing rabbits with a synthetic peptide corresponding to residues surrounding Glu31 of human STAG2. The antibody is purified via protein A and peptide affinity chromatography and is supplied in a formulation containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, and 50% glycerol.

  • Validated for Western blotting and immunoprecipitation applications.
  • Species reactivity includes human, mouse, rat, and monkey.
  • Target specificity ensures no cross-reactivity with other STAG proteins such as STAG1 or STAG3.
  • Supplied at a concentration of 49.3 μg/mL.
  • Recommended dilution for Western blotting is 1:1,000 and 1:100 for immunoprecipitation.
  • Storage at −20°C is required; do not aliquot.

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.