Learn More
Cell Signaling Technology T-bet/TBX21 (V365) Antibody, Polyclonal, Rabbit, 100 μl
SDP
Catalog No. 50195315
Encompass
T-bet/TBX21 (V365) Antibody is a polyclonal antibody designed to detect endogenous levels of total T-bet protein. It is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Val365 of the human T-bet/TBX21 protein and purified via protein A and peptide affinity chromatography. This antibody is suitable for applications such as Western blotting and immunoprecipitation.
  • Validated for human species reactivity
  • Targeted to endogenous protein levels
  • Purified by protein A and peptide affinity chromatography
  • Recommended dilution for Western blotting is 1:1,000
  • Recommended dilution for immunoprecipitation is 1:50
Catalog No. 50-195-315 Supplier Cell Signaling Technology Supplier No. 5214S
T-bet/TBX21 (V365) Antibody is a polyclonal antibody designed to detect endogenous levels of total T-bet protein. It is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Val365 of the human T-bet/TBX21 protein and purified via protein A and peptide affinity chromatography. This antibody is suitable for applications such as Western blotting and immunoprecipitation.
  • Validated for human species reactivity
  • Targeted to endogenous protein levels
  • Purified by protein A and peptide affinity chromatography
  • Recommended dilution for Western blotting is 1:1,000
  • Recommended dilution for immunoprecipitation is 1:50