Learn More
Bio X Cell Hamster IgG, Unconjugated Purity 95, 25 MG
Supplier: Bio X Cell BE0087R025MG
This polyclonal Syrian hamster IgG is purified from Syrian hamster serum and is unconjugated. With a purity of ≥95% and a molecular weight of 150 kDa, it serves as a non-reactive control IgG for Syrian hamster antibodies in various in vivo and in vitro applications.
- Isotype: Syrian hamster IgG
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
- Endotoxin level: ≤1EU/mg (≤0.001EU/µg)
- Sterility: 0.2 µm filtration
- Purification method: Protein G
- Store at stock concentration at 4°C; do not freeze
- Suitable for in vivo and in vitro applications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.