Learn More
Vector Laboratories POLYCLONAL RABBIT ANTI HU 50UL
SDP
Catalog No. NC3255259
Encompass
IL-31R-Beta antibody LS-B11477 is an unconjugated rabbit polyclonal antibody to human IL-31R-Beta OSMR /aa612-661/. Validated for IHC and WB. For Research Use Only.
Catalog No. NC3255259 Supplier Vector Laboratories Supplier No. LSB1147750
IL-31R-Beta antibody LS-B11477 is an unconjugated rabbit polyclonal antibody to human IL-31R-Beta OSMR /aa612-661/. Validated for IHC and WB. For Research Use Only.