Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PolyCo™ PolyWear™ Embossed Disposable Gowns, Open Back

Catalog No. 19125561
Encompass_Preferred
Change view
Click to view available options
Packaging Type:
Bulk Packaged
Dispenser Boxed, Individually Bagged
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type
19-125-561 Bulk Packaged
19-125-560 Dispenser Boxed, Individually Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-125-561 Supplier PolyCo™ Supplier No. 11520
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Large blue gowns feature easy tear-away for quick gown removal and safe disposal.

  • In compliance with FDA requirements and tested in accordance with ASTM standards including breaking load, tensile strength and elongation
  • Successfully tested for liquid and aerosol blood resistance under ASTM F 1670 Synthetic Blood Penetration Resistance and ASTM F 1671 Bacteriophage Penetration Resistance
  • In compliance with OSHA bloodborne pathogen standards (standard number 1910.1030)
  • AAMI levels 1 & 2 barrier protection
  • Superior physical properties over imports
  • Easy tear-away feature for quick gown removal and safe disposal
  • Contoured thumb loops secure gown sleeve for added protection
  • Redirects stress to the film material not the thermal seals to minimize breakage potential
  • Full-length open back affords better comfort and shoulder coverage
  • Provides liquid and aerosol blood resistance
  • Lightweight, non-linting construction

Order Info

Available in Bulk or Dispenser Boxed and individually bagged.

Specifications

Packaging Type Bulk Packaged
Size Large
For Use With (Application) Hospitals, Health Clinics, Laboratories
Gender Unisex
No. of Pockets None
Product Type Isolation Gown
Material Polyethylene Film
Color Blue
Closure Type Waist Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Thumb Loop
Finish Embossed
Model 11520
Certifications/Compliance ASTM F1670, ASTM F1671, AAMI Level 1 and 2, OSHA 29, CFR Part 1910.1030
Autoclavable Not Autoclavable
No. per Pack 100/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.