Learn More
PolyCo™ VR™ Protective Sleeves
Catalog No. 19125501
Change view
Click to view available options
Thickness:
4 mil
6 mil
Size:
Shoulder Length
Standard
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thickness Size
19-125-501 4 mil Standard
19-125-504 6 mil Standard
19-125-502 4 mil Standard
19-125-505 6 mil Standard
19-125-503 6 mil Standard
19-125-506 4 mil Shoulder Length
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-125-501 Supplier PolyCo™ Supplier No. 41802
Add to Cart
Add to Cart
This item is not returnable. View return policy
Easy to clean and reuse, VR sleeves are lightweight yet more durable than vinyl.
  • Waterproof - barrier protection
  • AAMI Class 2 chemical splash protection
  • Ideal for flexibility
  • Strong, natural rubber elastic cuffs
  • Textured inside surface minimizes cling and enhances ventilation
  • Smooth outside surface for ease of cleaning
  • Available in different colors and thicknesses
  • Made in USA
Specifications
Size Standard
Material VR™ Polyolefin
Disposable Reusable
Cuff Style Elastic
Finish Textured
Model 41802
Thickness 4 mil
For Use With (Application) Food Processing, Critical and Controlled Environments, Healthcare, Manufacturing
Certifications/Compliance 21 CFR 175.300
Length (English) 18 in.
Length (Metric) 45.7 cm