Learn More
PolyCo™ VR™ Protective Sleeves
Catalog No. 19125506
Encompass_Preferred
Change view
Click to view available options
Thickness:
4 mil
6 mil
Size:
Shoulder Length
Standard
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thickness Size
19-125-506 4 mil Shoulder Length
19-125-501 4 mil Standard
19-125-504 6 mil Standard
19-125-502 4 mil Standard
19-125-505 6 mil Standard
19-125-503 6 mil Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-125-506 Supplier PolyCo™ Supplier No. 41900
This item is not returnable. View return policy
Easy to clean and reuse, VR sleeves are lightweight yet more durable than vinyl.
  • Waterproof - barrier protection
  • AAMI Class 2 chemical splash protection
  • Ideal for flexibility
  • Strong, natural rubber elastic cuffs
  • Textured inside surface minimizes cling and enhances ventilation
  • Smooth outside surface for ease of cleaning
  • Available in different colors and thicknesses
  • Made in USA
Specifications
Size Shoulder Length
Material VR™ Polyolefin
Disposable Reusable
Cuff Style Elastic
Finish Textured
Model 41900
Thickness 4 mil
For Use With (Application) Food Processing, Critical and Controlled Environments, Healthcare, Manufacturing
Certifications/Compliance 21 CFR 175.300
Length (English) 24 in.
Length (Metric) 61 cm