Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PolyCo™ VR™ Rainwear Overalls

Catalog No. 19125551
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-125-551 Large
19-125-550 Medium
19-125-552 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-125-551 Supplier PolyCo™ Supplier No. 53524
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Closure Type Storm Flap with Heavy-duty Buckles
Color Yellow
Material PVC
Size Large
Construction 3-piece
For Use With (Application) Impervious to fats and Oils
Thickness 8 mil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.