Learn More
Cellucap Polycoated Propylene Gowns
Catalog No. 19042098 Shop All Cellucap Products
Change view
Click to view available options
Size:
Universal
X-Large
Cuff Style:
Elastic
Knit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Cuff Style
19-042-098 X-Large Elastic
18-022-979 Universal Knit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-042-098 Supplier Cellucap Supplier No. 434TXL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Polycoated gown with waist and neck ties provides fluid proof protection.
  • White fluid proof polycoated gown
  • Ties at neck and waste
  • Universal or extra large size
  • 5 bags of 10 gowns packaged 50 per case
Specifications
Size X-Large
Gender Unisex
No. of Pockets None
Product Type General Purpose Gown
Material Polypropylene
Color White
Closure Type Neck and Waist Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Model 434TXL
Sleeve Style Long Sleeve
Autoclavable Not Autoclavable