Learn More
Enzo Life Sciences Polycomb 2 polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS2910100
Encompass_Preferred
Isotype: IgG. Immunogen: Synthetic peptide corresponding to a portion of N-terminal human Polycomb 2, conjugated to KLH. Species reactivity: Human, Mouse. Applications: ELISA, IHC, WB. Purity: Affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide.
Catalog No. 50-201-2831