Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences NUNC™ Polycarbonate Membrane for Multidish 6, 3 µm pores
SDP

Catalog No. 5034416
Encompass_Preferred

Treated and tested for cell attachment. Easy cultivation of most cell types without matrix coating. Used in a varity of applications, including transport studies, toxicity tests, chemotaxis studies and electron microscopy. Non-toxic. Packed in sterile Nunclon™Δ treated multidishes. Packed in resealable bags. Non-pyrogenic. Tabs for better grip/handling.
Pore size: 3 µm
Porosity: <7.7 percent
Culture Area: 3.14 cm2
Working volume: 1.5 ml

Catalog No. 50-344-16 Supplier Electron Microscopy Sciences Supplier No. 7229623
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Treated and tested for cell attachment. Easy cultivation of most cell types without matrix coating. Used in a varity of applications, including transport studies, toxicity tests, chemotaxis studies and electron microscopy. Non-toxic. Packed in sterile Nunclon™Δ treated multidishes. Packed in resealable bags. Non-pyrogenic. Tabs for better grip/handling.
Pore size: 3 µm
Porosity: <7.7 percent
Culture Area: 3.14 cm2
Working volume: 1.5 ml

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.