Learn More
Chemglass Life Sciences Poly Crimp Seal, 11mm, Clear Cap, Starburst Septum
SDP
Catalog No. 501532589
Encompass_Preferred
Poly Crimp Seal, 11mm, Clear Cap, Starburst Septum. A crimp cap constructed of plastic that fits virtually any 11mm crimp sample vial. Use a standard crimping tool to securely crimp the Poly Crimp seal onto any crimp top vial. The Poly Crimp seal also permits an airtight seal by hand without crimping. Although the Poly Crimp seal can work with any crimp top vial, the tighter manufacturing tolerances and quality assurance program with the vials supplied by Chemglass certifies better performance when the cap is used with Chemglass supplied vials. Vials Compatible with Poly Crimp Seals: CV-1108-1232: Vial, 2.0mL, Clear, Snap Seal, 12 x 32mm, 11mm Crimp CV-1168-1232 : Vial, Standard Opening Crimp Top, Clear, 2.0mL, 12 x 32mm, 11mm Crimp CV-1185-1232 : Vial, Large Opening Crimp Top, Clear, 2.0mL, 12 x 32mm, 11mm Crimp CV-1200-1232 : Vial, Snap Ring, 2.0mL, Clear, 12 x 32mm, 11mm Crimp
Catalog No. 50-153-2589 Supplier Chemglass Life Sciences Supplier No. CV39100011
Poly Crimp Seal, 11mm, Clear Cap, Starburst Septum. A crimp cap constructed of plastic that fits virtually any 11mm crimp sample vial. Use a standard crimping tool to securely crimp the Poly Crimp seal onto any crimp top vial. The Poly Crimp seal also permits an airtight seal by hand without crimping. Although the Poly Crimp seal can work with any crimp top vial, the tighter manufacturing tolerances and quality assurance program with the vials supplied by Chemglass certifies better performance when the cap is used with Chemglass supplied vials. Vials Compatible with Poly Crimp Seals: CV-1108-1232: Vial, 2.0mL, Clear, Snap Seal, 12 x 32mm, 11mm Crimp CV-1168-1232 : Vial, Standard Opening Crimp Top, Clear, 2.0mL, 12 x 32mm, 11mm Crimp CV-1185-1232 : Vial, Large Opening Crimp Top, Clear, 2.0mL, 12 x 32mm, 11mm Crimp CV-1200-1232 : Vial, Snap Ring, 2.0mL, Clear, 12 x 32mm, 11mm Crimp