Learn More
AdipoGen PolyD-lactide
SDP
Supplier:  AdipoGen CDXP0018M100
Encompass_Preferred
Chemical. CAS 106989-11-1. Formula C3H4O2n. MW 90000-124000. Synthetic. PolyD-lactide PDLA, a polymer of a stereospecific cyclic di-ester of lactic acid, is used in biomaterial research for the development of devices such as therapeutic drug delivery vessels. PolyD-lactide is used for the preparation of microparticles and resorbable polylactide scaffolds.
Catalog No. 50-256-0280