Learn More
AdipoGen Polydatin (1 g)
SDP
Supplier:  AdipoGen CDXP0596G001
Encompass_Preferred
Chemical. CAS: 65914-17-2. Formula: C20H22O8. MW: 390.38. Polydatin is a natural stilbene with antioxidant, anti-inflammatory, anticancer, neuroprotective, cardioprotective, anti-osteoporotic and antilipidemic properties. It has been shown to inhibit NF-kappa signaling pathway and the NLRP3 inflammasome activation. It is an apoptosis and autophagy inducer, cell cycle inhibitor, activator of SIRT1 and also inhibits adipogenesis.
Catalog No. 50-205-4623