Learn More
Apexbio Technology LLC Polydatin 27208-80-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063287
Encompass_Preferred
Polydatin (CAS 27208-80-6) is a naturally occurring stilbenoid isolated primarily from the roots of Polygonum cuspidatum Structurally it represents the glucoside derivative of resveratrol exerting antioxidant and anti-inflammatory effects through modulation of oxidative stress-related pathways Research indicates that polydatin influences signaling cascades such as NF- B MAPK and Nrf2 contributing to its protective roles in cellular injury models including cardiovascular dysfunction neurodegeneration and inflammatory conditions Polydatin is soluble in methanol ethanol and acetone and stability is maintained by storage under sealed cool dry conditions
Catalog No. 50-000-63287 Supplier Apexbio Technology LLC Supplier No. N17835.1
Polydatin (CAS 27208-80-6) is a naturally occurring stilbenoid isolated primarily from the roots of Polygonum cuspidatum Structurally it represents the glucoside derivative of resveratrol exerting antioxidant and anti-inflammatory effects through modulation of oxidative stress-related pathways Research indicates that polydatin influences signaling cascades such as NF- B MAPK and Nrf2 contributing to its protective roles in cellular injury models including cardiovascular dysfunction neurodegeneration and inflammatory conditions Polydatin is soluble in methanol ethanol and acetone and stability is maintained by storage under sealed cool dry conditions