Learn More
Medchemexpress LLC Polydextrose | 68424-04-4 | MFCD30176547 | 95.3% | 2,000 Da (weight-average) | (C6H10O5)n | 25 G
SDP
Catalog No. 5000283387
Encompass_Preferred
Polydextrose is a synthetic, water-soluble polysaccharide used as a prebiotic and research reagent. It is commonly applied in studies of lipid and glucose metabolism, including hyperlipidemia and type 2 diabetes models, and as a soluble dietary fiber in formulation research. The material typically has a weight-average molecular weight of about 2,000 Da and is supplied at approximately 95.3% purity for laboratory use.
  • Water-soluble dietary fiber and prebiotic.
  • Supports research on lipid and glucose metabolism.
  • Broad molecular weight distribution with ~2,000 Da average.
  • High purity suitable for analytical and biological studies.
  • Stable under typical laboratory handling and storage conditions.
Catalog No. 50-002-83387 Supplier Medchemexpress LLC Supplier No. HYW13432625G
Polydextrose is a synthetic, water-soluble polysaccharide used as a prebiotic and research reagent. It is commonly applied in studies of lipid and glucose metabolism, including hyperlipidemia and type 2 diabetes models, and as a soluble dietary fiber in formulation research. The material typically has a weight-average molecular weight of about 2,000 Da and is supplied at approximately 95.3% purity for laboratory use.
  • Water-soluble dietary fiber and prebiotic.
  • Supports research on lipid and glucose metabolism.
  • Broad molecular weight distribution with ~2,000 Da average.
  • High purity suitable for analytical and biological studies.
  • Stable under typical laboratory handling and storage conditions.