Learn More
Medchemexpress LLC Polydextrose | 68424-04-4 | MFCD30176547 | 95.3% | 342.3 g/mol | C12H22O11 | 250 G
SDP
Catalog No. 5000283337
Encompass_Preferred
Polydextrose is a water-soluble, synthetic polymeric carbohydrate used as an orally active prebiotic in biomedical and metabolic research, including studies on gut microbiota, cholesterol metabolism, and glycemic control.
  • Water-soluble polymeric carbohydrate.
  • Orally active prebiotic for microbiota research.
  • Reported to reduce cholesterol and modulate blood glucose.
  • Typical assay purity 95.3%.
  • Supplied as a powdered product in laboratory package sizes up to 500 g.
  • CAS number 68424-04-4; empirical formula C12H22O11.
Catalog No. 50-002-83337 Supplier Medchemexpress LLC Supplier No. HYW134326250G
Polydextrose is a water-soluble, synthetic polymeric carbohydrate used as an orally active prebiotic in biomedical and metabolic research, including studies on gut microbiota, cholesterol metabolism, and glycemic control.
  • Water-soluble polymeric carbohydrate.
  • Orally active prebiotic for microbiota research.
  • Reported to reduce cholesterol and modulate blood glucose.
  • Typical assay purity 95.3%.
  • Supplied as a powdered product in laboratory package sizes up to 500 g.
  • CAS number 68424-04-4; empirical formula C12H22O11.