Learn More
Medchemexpress LLC Polydextrose | 68424-04-4 | MFCD30176547 | 95.3% | No single molecular weight (polymeric; variable) | (C6H10O5)n | 500 G
SDP
Catalog No. 5000283293
Encompass_Preferred
Polydextrose is a water-soluble, randomly bonded glucose polymer used as a prebiotic research reagent. It supports growth of beneficial probiotic strains and has been studied for effects on cholesterol and glycemic control, making it useful for investigations in metabolic and gastrointestinal research settings.
  • Water-soluble polymeric prebiotic suitable for research use.
  • Promotes growth of probiotic strains such as Pediococcus pentosaceus.
  • Studied for cholesterol-lowering and blood sugar-modulating effects.
  • Supplied as a powder for easy handling and formulation.
  • Typical listed purity approximately 95.3% (batch dependent).
Catalog No. 50-002-83293 Supplier Medchemexpress LLC Supplier No. HYW134326500G
Polydextrose is a water-soluble, randomly bonded glucose polymer used as a prebiotic research reagent. It supports growth of beneficial probiotic strains and has been studied for effects on cholesterol and glycemic control, making it useful for investigations in metabolic and gastrointestinal research settings.
  • Water-soluble polymeric prebiotic suitable for research use.
  • Promotes growth of probiotic strains such as Pediococcus pentosaceus.
  • Studied for cholesterol-lowering and blood sugar-modulating effects.
  • Supplied as a powder for easy handling and formulation.
  • Typical listed purity approximately 95.3% (batch dependent).